Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsnax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022262.3 | NP_071598.1 | 4 - 5 | 453 | 98 | |
GenBank mRNA | AF262357.1 | - | 4 - 5 | 524 | 98 | |
BC081715.1 | - | 4 - 5 | 453 | 7130295 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |