Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011927.1 | NP_001011927.1 | 2 - 3 | 380 | 73 | |
GenBank mRNA | BC088462.1 | - | 2 - 3 | 380 | 7127604 | 73 |
FQ213716.1 | - | 2 - 3 | 396 | 73 | ||
FQ215412.1 | - | 1 - 2 | 115 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |