Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC058481.1 | - | 1 - 2 | 69 | 6921522 | 91 |
FQ211079.1 | - | 1 - 2 | 58 | 91 | ||
FQ215547.1 | - | 1 - 2 | 94 | 91 | ||
FQ221561.1 | - | 1 - 2 | 63 | 91 | ||
FQ225602.1 | - | 1 - 2 | 65 | 91 | ||
FQ227282.1 | - | 1 - 2 | 65 | 91 | ||
FQ227580.1 | - | 1 - 2 | 56 | 91 | ||
FQ233937.1 | - | 1 - 2 | 65 | 91 | ||
FQ234358.1 | - | 1 - 2 | 94 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |