Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081634.1 | NP_001075103.1 | 7 - 8 | 1213 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |