Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007755.1 | NP_001007756.1 | 1 - 2 | 65 | 80 | |
XM_008767277.1 | XP_008765499.1 | 1 - 2 | 148 | 80 | ||
GenBank mRNA | BC079358.1 | - | 1 - 2 | 65 | 7108615 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |