Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108230.1 | NP_001101700.1 | 1 - 2 | 61 | 60 | |
XM_006245396.2 | XP_006245458.2 | 2 - 3 | 4157 | 60 | ||
XM_006245397.2 | XP_006245459.2 | 2 - 3 | 4157 | 60 | ||
XM_006245398.2 | XP_006245460.2 | 2 - 3 | 4156 | 60 | ||
XM_006245400.2 | XP_006245462.2 | 2 - 3 | 4156 | 60 | ||
XM_006245401.2 | XP_006245463.2 | 2 - 3 | 4157 | 60 | ||
XM_006245402.2 | XP_006245464.2 | 2 - 3 | 4157 | 60 | ||
XM_006245403.2 | XP_006245465.2 | 2 - 3 | 4156 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |