Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108810.1 | NP_001102280.1 | 3 - 4 | 410 | 86 | |
GenBank mRNA | BC167048.1 | - | 3 - 4 | 281 | 9100546 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |