Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135667.1 | NP_001129139.1 | 5 - 6 | 625 | 71 | |
XM_008772828.1 | XP_008771050.1 | 5 - 6 | 836 | 71 | ||
XM_008772829.1 | XP_008771051.1 | 5 - 6 | 770 | 71 | ||
XM_008772830.1 | XP_008771052.1 | 5 - 6 | 803 | 71 | ||
XM_008772831.1 | XP_008771053.1 | 3 - 4 | 401 | 71 | ||
XM_008772832.1 | XP_008771054.1 | 3 - 4 | 420 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |