Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135667.1 | NP_001129139.1 | 9 - 10 | 1036 | 129 | |
XM_008772828.1 | XP_008771050.1 | 9 - 10 | 1247 | 129 | ||
XM_008772829.1 | XP_008771051.1 | 9 - 10 | 1181 | 129 | ||
XM_008772830.1 | XP_008771052.1 | 9 - 10 | 1214 | 129 | ||
XM_008772831.1 | XP_008771053.1 | 7 - 8 | 812 | 129 | ||
XM_008772832.1 | XP_008771054.1 | 7 - 8 | 831 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |