Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212538.1 | NP_997703.1 | 6 - 7 | 811 | 72 | |
XM_006256117.2 | XP_006256179.1 | 6 - 7 | 1065 | 72 | ||
XM_008772723.1 | XP_008770945.1 | 6 - 7 | 1066 | 72 | ||
GenBank mRNA | BC083843.1 | - | 6 - 7 | 752 | 7114670 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |