Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053840.2 | NP_446292.2 | 1398 | 107 | ||
GenBank mRNA | BC078768.1 | - | 1398 | 7133071 | 107 | |
M33821.1 | - | 1497 | 107 | |||
M33822.1 | - | 1483 | 107 | |||
X15443.1 | - | 1404 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |