Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001077030.3 | XP_001077030.1 | 2 - 3 | 528 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |