Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013073.1 | NP_001013091.1 | 9 - 10 | 1662 | 84 | |
XM_006256185.2 | XP_006256247.1 | 9 - 10 | 1688 | 84 | ||
GenBank mRNA | BC087068.1 | - | 9 - 10 | 1662 | 7133235 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |