Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212491.2 | NP_997656.1 | 10 - 11 | 1162 | 152 | |
GenBank mRNA | BC085934.1 | - | 10 - 11 | 1162 | 7112671 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |