Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008885.2 | NP_001008885.1 | 1 - 2 | 724 | 91 | |
NM_001164744.1 | NP_001158216.1 | 2 - 3 | 465 | 91 | ||
GenBank mRNA | BC079141.1 | - | 1 - 2 | 724 | 7127902 | 91 |
DY547565.1 | - | 2 - 3 | 465 | 91 | ||
FQ212335.1 | - | 2 - 3 | 432 | 91 | ||
FQ212833.1 | - | 1 - 2 | 752 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |