Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 25 - 26 | 7489 | 136 | |
XM_006256359.2 | XP_006256421.1 | 25 - 26 | 7543 | 136 | ||
XM_006256360.2 | XP_006256422.1 | 24 - 25 | 7339 | 136 | ||
XM_006256361.2 | XP_006256423.1 | 25 - 26 | 7566 | 136 | ||
XM_006256362.2 | XP_006256424.1 | 25 - 26 | 7458 | 136 | ||
XM_006256363.1 | XP_006256425.1 | 21 - 22 | 6900 | 136 | ||
XM_006256364.1 | XP_006256426.1 | 25 - 26 | 7452 | 136 | ||
XM_006256365.1 | XP_006256427.1 | 20 - 21 | 6758 | 136 | ||
GenBank mRNA | AF202265.1 | - | 2 - 3 | 281 | 136 | |
AY327507.1 | - | 5 - 6 | 677 | 136 | ||
FQ235173.1 | - | 3 - 4 | 417 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |