Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025144.1 | NP_001020315.1 | 8 - 9 | 1106 | 108 | |
GenBank mRNA | BC079112.1 | - | 8 - 9 | 1106 | 7125748 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |