Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109589.2 | NP_001103059.1 | 2 - 3 | 135 | 78 | |
XM_006256342.2 | XP_006256404.1 | 2 - 3 | 565 | 78 | ||
XM_008772925.1 | XP_008771147.1 | 2 - 3 | 570 | 78 | ||
XM_008772926.1 | XP_008771148.1 | 2 - 3 | 571 | 78 | ||
GenBank mRNA | BC167029.1 | - | 2 - 3 | 135 | 8368465 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |