Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107622.1 | NP_001101092.1 | 10 - 11 | 2705 | 99 | |
XM_008772882.1 | XP_008771104.1 | 11 - 12 | 4738 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |