Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053845.1 | NP_446297.1 | 8 - 9 | 948 | 126 | |
GenBank mRNA | BC078767.1 | - | 8 - 9 | 938 | 7130296 | 126 |
FQ209442.1 | - | 8 - 9 | 926 | 126 | ||
M97662.1 | - | 8 - 9 | 948 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |