Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772939.1 | XP_008771161.1 | 5 - 6 | 7540 | 110 | |
XM_008772940.1 | XP_008771162.1 | 5 - 6 | 7540 | 110 | ||
GenBank mRNA | FQ212141.1 | - | 2 - 3 | 287 | 110 | |
FQ212194.1 | - | 1304 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |