Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107624.1 | NP_001101094.1 | 1208 | 123 | ||
GenBank mRNA | FQ229049.1 | - | 3 - 4 | 311 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |