Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-Db2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164826.1 | NP_001158298.1 | 2 - 3 | 370 | 129 | |
XM_006255911.2 | XP_006255973.1 | 2 - 3 | 511 | 129 | ||
GenBank mRNA | KC222923.1 | - | 2 - 3 | 370 | 129 | |
KC222924.1 | - | 2 - 3 | 370 | 129 | ||
KC222925.1 | - | 2 - 3 | 370 | 129 | ||
KC222926.1 | - | 2 - 3 | 370 | 129 | ||
KC222927.1 | - | 2 - 3 | 370 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |