Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025016.1 | NP_001020187.1 | 1 - 2 | 141 | 84 | |
GenBank mRNA | BC082031.1 | - | 1 - 2 | 141 | 7130142 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |