Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031831.1 | NP_114019.1 | 6 - 7 | 3591 | 161 | |
XM_006251608.1 | XP_006251670.1 | 4 - 5 | 1368 | 161 | ||
XM_006251609.2 | XP_006251671.1 | 4 - 5 | 1078 | 161 | ||
XM_008770468.1 | XP_008768690.1 | 5 - 6 | 5178 | 161 | ||
GenBank mRNA | AF051335.1 | - | 4 - 5 | 503 | 161 | |
AF132045.1 | - | 5 - 6 | 1687 | 161 | ||
AJ242961.1 | - | 6 - 7 | 3591 | 161 | ||
AJ242962.1 | - | 4 - 5 | 1118 | 161 | ||
AJ242963.1 | - | 4 - 5 | 564 | 161 | ||
AY164740.1 | - | 4 - 5 | 1118 | 161 | ||
AY164741.1 | - | 5 - 6 | 1687 | 161 | ||
BC070879.1 | - | 4 - 5 | 1303 | 161 | ||
BC097936.1 | - | 4 - 5 | 1123 | 161 | ||
BC161841.1 | - | 4 - 5 | 1124 | 161 | ||
FQ213608.1 | - | 2 - 3 | 163 | 161 | ||
FQ228713.1 | - | 3 - 4 | 301 | 161 | ||
FQ235341.1 | - | 3 - 4 | 264 | 161 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |