Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106759.2 | NP_001100229.1 | 2 - 3 | 690 | 110 | |
XM_006240572.2 | XP_006240634.1 | 2 - 3 | 690 | 110 | ||
XM_006240573.2 | XP_006240635.1 | 2 - 3 | 648 | 110 | ||
XM_006240574.2 | XP_006240636.1 | 2 - 3 | 3068 | 110 | ||
XM_006240575.2 | XP_006240637.1 | 2 - 3 | 794 | 110 | ||
XM_006240576.2 | XP_006240638.1 | 2 - 3 | 457 | 110 | ||
GenBank mRNA | BC166426.1 | - | 2 - 3 | 690 | 7100899 | 110 |
FQ230530.1 | - | 2 - 3 | 709 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |