Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012689.1 | NP_036821.1 | 3 - 4 | 859 | 91 | |
XM_006227832.2 | XP_006227894.1 | 3 - 4 | 1459 | 91 | ||
XM_008758703.1 | XP_008756925.1 | 4 - 5 | 1507 | 91 | ||
XM_008758704.1 | XP_008756926.1 | 3 - 4 | 1431 | 91 | ||
GenBank mRNA | AB477039.1 | - | 3 - 4 | 865 | 91 | |
KJ746832.1 | - | 2 - 3 | 650 | 91 | ||
KJ746833.1 | - | 2 - 3 | 650 | 91 | ||
X61098.1 | - | 2 - 3 | 723 | 91 | ||
Y00102.1 | - | 3 - 4 | 859 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Rattus norvegicus Probe spans exons |