Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108815.1 | NP_001102285.1 | 1 | 91 | 68 | |
GenBank mRNA | BC079470.1 | - | 1 | 69 | 68 | |
BC158792.1 | - | 1 | 110 | 7312115 | 68 | |
EF688600.1 | - | 1 | 69 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Amplicon spans exons and probe does not span exons |