Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010948.1 | NP_001010948.1 | 2 - 3 | 180 | 110 | |
GenBank mRNA | AY825929.1 | - | 2 - 3 | 180 | 110 | |
FQ211514.1 | - | 372 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |