Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010948.1 | NP_001010948.1 | 8 - 9 | 723 | 75 | |
GenBank mRNA | AY825929.1 | - | 8 - 9 | 723 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |