Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133415.1 | NP_596906.1 | 5 - 6 | 428 | 94 | |
XM_006255703.2 | XP_006255765.1 | 3 - 4 | 1494 | 94 | ||
GenBank mRNA | AF193757.1 | - | 5 - 6 | 428 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |