Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005381.2 | NP_001005381.2 | 5 - 6 | 460 | 62 | |
XM_008772490.1 | XP_008770712.1 | 246 | 62 | |||
GenBank mRNA | BC079094.1 | - | 5 - 6 | 431 | 7123989 | 62 |
CB582878.1 | - | 5 - 6 | 460 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |