Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox4i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017202.1 | NP_058898.1 | 3 - 4 | 336 | 99 | |
GenBank mRNA | BC084719.1 | - | 2 - 3 | 252 | 7317602 | 99 |
FQ214265.1 | - | 3 - 4 | 345 | 99 | ||
FQ217878.1 | - | 3 - 4 | 304 | 99 | ||
FQ223981.1 | - | 3 - 4 | 298 | 99 | ||
FQ228805.1 | - | 3 - 4 | 286 | 99 | ||
X14209.1 | - | 3 - 4 | 318 | 99 | ||
X15029.1 | - | 2 - 3 | 263 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |