Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192009.1 | NP_001178938.1 | 3 - 4 | 522 | 93 | |
XM_006255721.2 | XP_006255783.1 | 3 - 4 | 2232 | 93 | ||
XM_006255722.2 | XP_006255784.1 | 4 - 5 | 1880 | 93 | ||
XM_008772644.1 | XP_008770866.1 | 3 - 4 | 2232 | 93 | ||
XM_008772645.1 | XP_008770867.1 | 3 - 4 | 2232 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Rattus norvegicus 1700 Probe spans exons |