Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem184c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178330.1 | NP_847900.1 | 7 - 8 | 1234 | 119 | |
XM_006255422.2 | XP_006255484.1 | 8 - 9 | 946 | 119 | ||
XM_008772475.1 | XP_008770697.1 | 8 - 9 | 1845 | 119 | ||
GenBank mRNA | AY228474.1 | - | 7 - 8 | 1234 | 119 | |
BC089112.1 | - | 7 - 8 | 1244 | 7316181 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |