Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129881.2 | NP_001123353.1 | 3 - 4 | 264 | 102 | |
GenBank mRNA | BC166883.1 | - | 3 - 4 | 264 | 6919704 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |