Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053516.2 | NP_445968.2 | 1 - 2 | 378 | 65 | |
XM_006255539.2 | XP_006255601.1 | 2 - 3 | 509 | 65 | ||
XM_006255542.2 | XP_006255604.1 | 2 - 3 | 509 | 65 | ||
XM_008772540.1 | XP_008770762.1 | 2 - 3 | 990 | 65 | ||
GenBank mRNA | U40627.1 | - | 1 - 2 | 378 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |