Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013890.1 | NP_001013912.1 | 3 - 4 | 268 | 99 | |
GenBank mRNA | BC078968.1 | - | 3 - 4 | 268 | 7111580 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |