Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017175.2 | NP_058871.2 | 5 - 6 | 936 | 78 | |
XM_006255245.2 | XP_006255307.1 | 5 - 6 | 1976 | 78 | ||
GenBank mRNA | BC061836.1 | - | 5 - 6 | 837 | 5599989 | 78 |
D26180.1 | - | 5 - 6 | 887 | 78 | ||
FQ222217.1 | - | 5 - 6 | 936 | 78 | ||
L35634.1 | - | 5 - 6 | 779 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |