Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011928.1 | NP_001011928.1 | 7 - 8 | 1000 | 87 | |
XM_008772487.1 | XP_008770709.1 | 7 - 8 | 1297 | 87 | ||
GenBank mRNA | BC088195.1 | - | 7 - 8 | 1000 | 7374282 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |