Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008366.1 | NP_001008367.1 | 6 - 7 | 578 | 139 | |
XM_006255686.2 | XP_006255748.1 | 6 - 7 | 692 | 139 | ||
XM_006255687.2 | XP_006255749.1 | 6 - 7 | 610 | 139 | ||
XM_006255689.2 | XP_006255751.1 | 7 - 8 | 914 | 139 | ||
XM_006255690.2 | XP_006255752.1 | 7 - 8 | 700 | 139 | ||
GenBank mRNA | BC085839.1 | - | 6 - 7 | 578 | 7135799 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |