Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054009.1 | NP_446461.1 | 3 - 4 | 235 | 132 | |
GenBank mRNA | AB020757.1 | - | 3 - 4 | 235 | 132 | |
FQ234836.1 | - | 3 - 4 | 247 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |