Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004088.3 | NP_001004088.3 | 12 - 13 | 1955 | 65 | |
XM_006255504.1 | XP_006255566.1 | 12 - 13 | 1937 | 65 | ||
GenBank mRNA | AY248698.1 | - | 11 - 12 | 1913 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |