Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100922.1 | NP_001094392.1 | 3 - 4 | 370 | 116 | |
XM_006255219.1 | XP_006255281.1 | 3 - 4 | 304 | 116 | ||
GenBank mRNA | AY375243.1 | - | 3 - 4 | 216 | 116 | |
BC129128.1 | - | 3 - 4 | 370 | 7314651 | 116 | |
X57986.1 | - | 3 - 4 | 280 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |