Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039030.1 | NP_001034119.1 | 2 - 3 | 118 | 107 | |
XM_006255779.2 | XP_006255841.1 | 2 - 3 | 700 | 107 | ||
GenBank mRNA | BC099759.1 | - | 2 - 3 | 118 | 7929646 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |