Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbndd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014156.1 | NP_001014178.2 | 1 - 2 | 229 | 71 | |
GenBank mRNA | BC088277.1 | - | 1 - 2 | 229 | 7381798 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |