Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbndd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014156.1 | NP_001014178.2 | 3 - 4 | 530 | 59 | |
GenBank mRNA | BC088277.1 | - | 3 - 4 | 530 | 7381798 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |