Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtss1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191558.1 | NP_001178487.1 | 7 - 8 | 618 | 130 | |
XM_008772512.1 | XP_008770734.1 | 8 - 9 | 633 | 130 | ||
XM_008772513.1 | XP_008770735.1 | 7 - 8 | 583 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |