Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008553.1 | NP_001008553.1 | 2 - 3 | 89 | ||
XM_006255601.2 | XP_006255663.1 | 2 - 3 | 462 | 89 | ||
XM_008772529.1 | XP_008770751.1 | 2 - 3 | 462 | 89 | ||
GenBank mRNA | X80778.1 | - | 2 - 3 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |