Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012164.1 | NP_001012164.1 | 18 - 19 | 2379 | 55 | |
XM_006255278.2 | XP_006255340.1 | 18 - 19 | 3046 | 55 | ||
XM_006255279.2 | XP_006255341.1 | 17 - 18 | 2911 | 55 | ||
XM_006255280.1 | XP_006255342.1 | 19 - 20 | 2628 | 55 | ||
XM_006255281.2 | XP_006255343.1 | 16 - 17 | 2764 | 55 | ||
GenBank mRNA | BC083857.1 | - | 18 - 19 | 2379 | 7125718 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |